Assay Details
Target Gene Details
Entrez Gene ID: | 381022 |
Gene Name: | lysine (K)-specific methyltransferase 2D |
Gene Aliases: |
ALR, BC032281, BC058659, C430014K11Rik, KMT2B, Mll2, Mll4 |
Location: |
Chr.15:98831669-98871205 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 43 - Exon 43 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kmt2d | NM_001033276.3 | NP_001028448.3 | ||
| BC058659.1 | AAH58659.1 |

Genomic Map