Assay Details
Target Gene Details
Entrez Gene ID: | 67747 |
Gene Name: | RIB43A domain with coiled-coils 2 |
Gene Aliases: |
4930579A10Rik, Trib |
Location: |
Chr.15:85132060-85144569 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ribc2 | NM_026357.2 | 5 | 982 | NP_080633.2 |
| AK006801.1 | 6 | 1091 | ||
| AK016311.1 | 5 | 983 | BAB30190.2 | |
| AK017028.1 | 5 | 1001 | ||
| BC112373.1 | 5 | 907 | AAI12374.1 |
More Information
Set Membership: |
|

Genomic Map