Assay Details
Target Gene Details
Entrez Gene ID: | 406220 |
Gene Name: | keratin 77 |
Gene Aliases: |
4732484G22Rik, K1emb, Krt239, mK1b |
Location: |
Chr.15:101858731-101869706 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt77 | NM_001003667.1 | NP_001003667.1 | ||
| AK132513.1 | ||||
| BC107395.2 | ||||
| BC107396.2 | ||||
| BC141259.1 |
More Information
Set Membership: |
|

Genomic Map