Assay Details
Target Gene Details
Entrez Gene ID: | 71474 |
Gene Name: | protein phosphatase 6, regulatory subunit 2 |
Gene Aliases: |
1110033O10Rik, 8430411H09Rik, B230107H12Rik, Pp6r2, Saps2 |
Location: |
Chr.15:89211521-89287015 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp6r2 | NM_026813.1 | NP_081089.1 | ||
| NM_027805.2 | 8 | 1276 | NP_082081.1 | |
| AK018403.1 | 8 | 1195 | ||
| AK084684.1 | 8 | 1277 | BAC39251.1 | |
| AK173000.1 | BAD32278.1 | |||
| BC024879.1 | AAH24879.1 |
More Information
Set Membership: |
|

Genomic Map