Assay Details
Target Gene Details
Entrez Gene ID: | 60597 |
Gene Name: | mitogen-activated protein kinase 8 interacting protein 2 |
Gene Aliases: |
3230402N03Rik, AI847694, AW049958, IB2, Jip2, R75148 |
Location: |
Chr.15:89453911-89462447 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk8ip2 | NM_021921.3 | NP_068740.3 | ||
| AF220195.1 | AAL50331.1 | |||
| AF310135.1 | AAG31800.1 | |||
| AK014339.1 | ||||
| AK158968.1 | BAE34748.1 | |||
| BC029704.1 | AAH29704.1 | |||
| CA317486.1 |
More Information
Set Membership: |
|

Genomic Map