Assay Details
Target Gene Details
Entrez Gene ID: | 68070 |
Gene Name: | PDZ domain containing 2 |
Gene Aliases: |
4930537L06Rik, A930022H17Rik, Gm82, Pdzk3, mKIAA0300 |
Location: |
Chr.15:12357054-12739973 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdzd2 | NM_001081064.1 | NP_001074533.1 | ||
| AK020883.1 | BAB32241.3 | |||
| AK162516.1 | BAE36953.1 | |||
| BC115886.1 |

Genomic Map