Assay Details
Target Gene Details
Entrez Gene ID: | 20788 |
Gene Name: | sterol regulatory element binding factor 2 |
Gene Aliases: |
AI608257, SREBP-2, SREBP2, SREBP2gc, bHLHd2, lop13, nuc |
Location: |
Chr.15:82147269-82205379 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srebf2 | NM_033218.1 | NP_150087.1 | ||
| AF374267.1 | AAK54763.1 | |||
| AK155857.1 | BAE33463.1 | |||
| AK172219.1 | ||||
| AK191062.1 | ||||
| CB228109.2 |

Genomic Map