Assay Details
Target Gene Details
Entrez Gene ID: | 223455 |
Gene Name: | membrane-associated ring finger (C3HC4) 6 |
Gene Aliases: |
3830408G03, F830029L24Rik, mKIAA0597 |
Location: |
Chr.15:31455894-31531048 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| March6 | NM_172606.2 | NP_766194.2 | ||
| AK028353.1 | ||||
| AK042980.1 | BAC31427.1 | |||
| AK089827.1 | BAC40971.1 | |||
| AK129169.1 | BAC97979.1 | |||
| AK132461.1 | ||||
| AK218077.1 | ||||
| BC037454.1 | AAH37454.1 | |||
| BC048816.1 | AAH48816.1 | |||
| BC059190.1 |
More Information
Set Membership: |
|

Genomic Map