Assay Details
Target Gene Details
Entrez Gene ID: | 19387 |
Gene Name: | RAN GTPase activating protein 1 |
Gene Aliases: |
C79654, Fug1, mKIAA1835 |
Location: |
Chr.15:81704248-81729919 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rangap1 | NM_001146174.1 | NP_001139646.1 | ||
| NM_011241.4 | NP_035371.4 | |||
| AK042930.1 | ||||
| AK088768.1 | BAC40559.1 | |||
| AK129452.1 | BAC98262.1 | |||
| AK133905.1 | BAE21919.1 | |||
| AK145459.1 | BAE26449.1 | |||
| AK146670.1 | BAE27347.1 | |||
| AK159009.1 | BAE34767.1 | |||
| AK159525.1 | BAE35154.1 | |||
| AK159930.1 | ||||
| AK179009.1 | ||||
| AK205795.1 | ||||
| AK213698.1 | ||||
| AK214243.1 | ||||
| BC014855.1 | AAH14855.1 | |||
| BC052862.1 | AAH52862.1 | |||
| BC066213.1 | AAH66213.1 | |||
| BC071200.1 | AAH71200.1 | |||
| U08110.1 | AAA17681.1 |
More Information
Set Membership: |
|

Genomic Map