Assay Details
Target Gene Details
Entrez Gene ID: | 54381 |
Gene Name: | carboxypeptidase Q |
Gene Aliases: |
1190003P12Rik, 2610034C17Rik, Hls2, Lal-1, Pgcp |
Location: |
Chr.15:33083129-33594552 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpq | NM_018755.2 | 5 | 1050 | NP_061225.2 |
| NM_176073.4 | 4 | 960 | NP_788262.1 | |
| AF009513.1 | 4 | 841 | AAC17945.1 | |
| AF107835.1 | 4 | 928 | AAD43215.1 | |
| AK032972.1 | 4 | 825 | BAC28105.1 | |
| AK075686.1 | 4 | 891 | BAC35891.1 | |
| AK135096.1 | 4 | 947 | BAE22419.1 | |
| AK137164.1 | 4 | 834 | BAE23259.1 | |
| AK152139.1 | 5 | 928 | BAE30978.1 | |
| AK152145.1 | 5 | 928 | BAE30982.1 | |
| AK154361.1 | 4 | 570 | BAE32537.1 | |
| BC037067.1 | 4 | 807 | AAH37067.1 |
More Information
Set Membership: |
|

Genomic Map