Assay Details
Target Gene Details
Entrez Gene ID: | 75772 |
Gene Name: | patatin-like phospholipase domain containing 5 |
Gene Aliases: |
4833426H19Rik, GS2L |
Location: |
Chr.15:84112356-84127992 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnpla5 | NM_029427.1 | NP_083703.1 | ||
| AK014771.1 | BAB29543.1 | |||
| BC109360.2 | AAI09361.1 | |||
| BC109361.2 | AAI09362.1 |
More Information
Set Membership: |
|

Genomic Map