Assay Details
Target Gene Details
Entrez Gene ID: | 14000 |
Gene Name: | drosha, ribonuclease type III |
Gene Aliases: |
1110013A17Rik, AI874853, Etohi2, Rn3, Rnasen |
Location: |
Chr.15:12824804-12935291 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Drosha | NM_001130149.1 | NP_001123621.1 | ||
| NM_026799.3 | NP_081075.3 | |||
| AF533013.1 | AAO65849.1 | |||
| AK077549.1 | ||||
| AK144147.1 | BAE25729.1 | |||
| AK148640.1 | BAE28629.1 | |||
| BC050057.1 | AAH50057.1 | |||
| BC055696.1 | AAH55696.1 | |||
| BC057687.1 | AAH57687.1 | |||
| BC060265.1 | AAH60265.1 | |||
| BC088999.1 | AAH88999.1 |
More Information
Set Membership: |
|

Genomic Map