Assay Details
Target Gene Details
Entrez Gene ID: | 20491 |
Gene Name: | src-like adaptor |
Gene Aliases: |
Slap, Slap-1 |
Location: |
Chr.15:66780819-66831829 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sla | NM_001029841.4 | 2 | 315 | NP_001025012.1 |
| NM_009192.3 | 3 | 584 | NP_033218.1 | |
| AJ131777.1 | 2 | 222 | CAB66139.1 | |
| AK036167.1 | 2 | 316 | BAC29328.1 | |
| AK037901.1 | 2 | 229 | BAC29896.1 | |
| AK041565.1 | 1 | 94 | BAC30988.1 | |
| AK137954.1 | 2 | 250 | BAE23522.1 | |
| AK163948.1 | 1 | 2276 | ||
| BC032922.1 | 2 | 120 | AAH32922.1 | |
| CD578683.1 | 3 | 401 | ||
| CT010282.1 | CAJ18490.1 | |||
| U29056.1 | AAA82756.1 |
Target Gene Details
Entrez Gene ID: | 21819 |
Gene Name: | thyroglobulin |
Gene Aliases: |
Tgn, cog |
Location: |
Chr.15:66670756-66850721 on Build GRCm38 |
Assay Gene Location: | Within Intron 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tg | NM_009375.2 | NP_033401.2 | ||
| AB332093.1 | ||||
| AF076186.1 | AAC32268.1 | |||
| AF076187.1 | AAC32269.1 | |||
| BC111467.1 | AAI11468.1 | |||
| U76389.1 | AAB53204.1 |
More Information
Set Membership: |
|

Genomic Map