Assay Details
Target Gene Details
Entrez Gene ID: | 64706 |
Gene Name: | signal peptide, CUB domain, EGF-like 1 |
Gene Aliases: |
7330410C13Rik, A630023E24Rik |
Location: |
Chr.15:83602583-83725049 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scube1 | NM_001271472.1 | NP_001258401.1 | ||
| NM_001271473.1 | NP_001258402.1 | |||
| NM_022723.3 | NP_073560.2 | |||
| AF276425.1 | AAG25939.1 | |||
| AK041590.1 | BAC30996.1 | |||
| BC066066.1 | AAH66066.1 | |||
| BC080278.1 | AAH80278.1 |
More Information
Set Membership: |
|

Genomic Map