Assay Details
Target Gene Details
Entrez Gene ID: | 93686 |
Gene Name: | RNA binding protein, fox-1 homolog (C. elegans) 2 |
Gene Aliases: |
Fbm2, Fxh, Hrnbp2, Rbm9 |
Location: |
Chr.15:77078990-77307053 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbfox2 | NM_001110827.2 | NP_001104297.1 | ||
| NM_001110828.2 | NP_001104298.1 | |||
| NM_001110829.2 | NP_001104299.1 | |||
| NM_001110830.2 | NP_001104300.1 | |||
| NM_001286417.1 | NP_001273346.1 | |||
| NM_001286418.1 | NP_001273347.1 | |||
| NM_001286419.1 | NP_001273348.1 | |||
| NM_053104.6 | NP_444334.3 | |||
| NM_175387.3 | NP_780596.1 | |||
| AF229055.1 | AAL71902.1 | |||
| AF387322.1 | AAK64287.1 | |||
| AK044929.1 | BAC32147.1 | |||
| AK077594.1 | BAC36885.1 | |||
| AK139555.1 | ||||
| AY659951.1 | AAV74334.1 | |||
| AY659952.1 | AAV74335.1 | |||
| AY659953.1 | AAV74336.1 | |||
| AY904025.1 | ||||
| BC002124.1 | ||||
| BC027263.1 | AAH27263.1 | |||
| DQ017388.1 | ||||
| DQ017389.1 |
More Information
Set Membership: |
|

Genomic Map