Assay Details
Target Gene Details
Entrez Gene ID: | 21411 |
Gene Name: | transcription factor 20 |
Gene Aliases: |
2810438H08Rik, SPBP, mKIAA0292 |
Location: |
Chr.15:82808447-82987867 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcf20 | NM_001114140.1 | NP_001107612.1 | ||
| NM_013836.3 | NP_038864.3 | |||
| AK122239.1 | BAC65521.1 | |||
| AK152165.1 | BAE30999.1 | |||
| AY007594.1 | AAG28929.1 | |||
| BC080855.1 | AAH80855.1 | |||
| BC138038.1 | ||||
| U20282.1 | AAA86495.1 |
More Information
Set Membership: |
|

Genomic Map