Assay Details
Target Gene Details
Entrez Gene ID: | 382985 |
Gene Name: | ribonucleotide reductase M2 B (TP53 inducible) |
Gene Aliases: |
p53R2 |
Location: |
Chr.15:37923952-37961108 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrm2b | NM_199476.1 | NP_955770.1 | ||
| AK138731.1 | BAE23760.1 | |||
| BC058103.1 | AAH58103.1 |
More Information
Set Membership: |
|

Genomic Map