Assay Details
Target Gene Details
Entrez Gene ID: | 11658 |
Gene Name: | activated leukocyte cell adhesion molecule |
Gene Aliases: |
AI853494, BEN, CD166, DM-GRASP, MuSC, SC1 |
Location: |
Chr.16:52248996-52453255 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alcam | NM_009655.2 | 14 | 2147 | NP_033785.1 |
| AB003026.1 | 14 | 1809 | BAD18062.1 | |
| AK030851.1 | 14 | 2148 | BAC27159.1 | |
| AK161090.1 | 1 | 61 | BAE36190.1 | |
| BC027280.1 | 14 | 1804 | AAH27280.1 | |
| L25274.1 | 9 | 939 | AAA37528.1 | |
| U95030.1 | 14 | 1615 | AAC06342.1 |

Genomic Map