Assay Details
Target Gene Details
Entrez Gene ID: | 68942 |
Gene Name: | charged multivesicular body protein 2B |
Gene Aliases: |
1190006E07Rik |
Location: |
Chr.16:65539133-65562697 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chmp2b | NM_026879.2 | NP_081155.1 | ||
| AK004506.1 | BAB23339.1 | |||
| AK084111.1 | BAC39118.1 | |||
| AK210184.1 | ||||
| AK216188.1 | ||||
| AK216245.1 | ||||
| AK217551.1 | ||||
| BC042626.1 | AAH42626.1 | |||
| BC055809.1 | AAH55809.1 |
More Information
Set Membership: |
|

Genomic Map