Assay Details
Target Gene Details
Entrez Gene ID: | 208650 |
Gene Name: | Casitas B-lineage lymphoma b |
Gene Aliases: |
AI429560, AI851073, Cbl-b |
Location: |
Chr.16:52031230-52208047 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cblb | NM_001033238.1 | NP_001028410.1 | ||
| AK147367.1 | ||||
| AK156295.1 | ||||
| AK161486.1 | BAE36418.1 | |||
| AK170640.1 | ||||
| AK171350.1 | ||||
| BC150934.1 | ||||
| BC150938.1 |
More Information
Set Membership: |
|

Genomic Map