Assay Details
Target Gene Details
Entrez Gene ID: | 12667 |
Gene Name: | chordin |
Gene Aliases: |
Chd |
Location: |
Chr.16:20731417-20742384 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrd | NM_001278041.1 | 2 | 248 | NP_001264970.1 |
| NM_009893.2 | NP_034023.1 | |||
| AF069501.1 | AAC68867.1 | |||
| AF096276.1 | AAD19895.1 | |||
| AK077460.1 | ||||
| AK149445.1 | BAE28881.1 | |||
| AK165339.1 | ||||
| BC145696.1 | 2 | 246 |
Target Gene Details
Entrez Gene ID: | 21832 |
Gene Name: | thrombopoietin |
Gene Aliases: |
Mgdf, Ml, Mpllg, Tpo |
Location: |
Chr.16:20724454-20734511 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thpo | NM_009379.3 | NP_033405.1 | ||
| AK047512.1 | BAC33078.1 |
More Information
Set Membership: |
|

Genomic Map