Assay Details
Target Gene Details
Entrez Gene ID: | 73261 |
Gene Name: | RIKEN cDNA 1700037C18 gene |
Gene Aliases: |
2810022L20Rik |
Location: |
Chr.16:3905798-3908690 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700037C18Rik | NM_028484.2 | 1 | 78 | NP_082760.2 |
| AK012792.1 | 1 | 49 | BAC25379.1 |
Target Gene Details
Entrez Gene ID: | 76779 |
Gene Name: | clusterin associated protein 1 |
Gene Aliases: |
2310030D15Rik, 2610111M03Rik |
Location: |
Chr.16:3907659-3941147 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cluap1 |

Genomic Map