Assay Details
Target Gene Details
Entrez Gene ID: | 56297 |
Gene Name: | ADP-ribosylation factor-like 6 |
Gene Aliases: |
1110018H24Rik, 2210411E14Rik, BBS3 |
Location: |
Chr.16:59613321-59639405 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arl6 | NM_019665.3 | NP_062639.3 | ||
| AB232697.1 | BAE79900.1 | |||
| AF031903.1 | AAC62194.1 | |||
| AK003778.1 | BAB22990.1 | |||
| AK152980.1 | BAE31631.1 | |||
| AK158832.1 | BAE34686.1 | |||
| AK160666.1 | BAE35949.1 | |||
| BC018497.1 | AAH18497.1 |
More Information
Set Membership: |
|

Genomic Map