Assay Details
Target Gene Details
Entrez Gene ID: | 67501 |
Gene Name: | coiled-coil domain containing 50 |
Gene Aliases: |
C3orf6 |
Location: |
Chr.16:27388765-27452218 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc50 | NM_001025615.3 | NP_001020786.1 | ||
| NM_001289436.1 | NP_001276365.1 | |||
| NM_026202.4 | NP_080478.2 | |||
| AJ534985.1 | CAD59434.1 | |||
| AK012190.1 | BAB28089.1 | |||
| AK016827.1 | BAB30453.1 | |||
| AK147820.1 | ||||
| AK151672.1 | BAE30598.1 | |||
| AK162434.1 | BAE36915.1 | |||
| AK165214.1 | BAE38081.1 | |||
| BC144895.1 | ||||
| BC145933.1 | ||||
| BC145935.1 | ||||
| CD553033.1 |

Genomic Map