Assay Details
Target Gene Details
Entrez Gene ID: | 67367 |
Gene Name: | PAX3 and PAX7 binding protein 1 |
Gene Aliases: |
1810007M14Rik, C21orf66, Gcfc, Gcfc1, Pax3/7bp |
Location: |
Chr.16:91014037-91044734 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paxbp1 | NM_026110.2 | 12 | 1992 | NP_080386.3 |
| AK007365.1 | 6 | 697 | BAB24988.2 | |
| AK011477.1 | 2 | 76 | BAB27645.2 | |
| AK079394.1 | 5 | 857 | ||
| AY033907.1 | 12 | 1777 | AAK68725.1 | |
| AY033908.1 | 12 | 1794 | AAK68726.1 | |
| BC014838.1 | 5 | 477 | ||
| BC027145.1 | 4 | 412 | AAH27145.1 |
More Information
Set Membership: |
|

Genomic Map