Assay Details
Target Gene Details
Entrez Gene ID: | 70790 |
Gene Name: | ubiquitin protein ligase E3 component n-recognin 5 |
Gene Aliases: |
Edd, Edd1 |
Location: |
Chr.15:37967328-38078854 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 50 - Exon 51 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubr5 | NM_001081359.3 | NP_001074828.2 | ||
| NM_001112721.2 | NP_001106192.1 | |||
| AK122398.1 | BAC65680.1 | |||
| AY550908.1 | AAT28194.1 | |||
| BC049162.1 | AAH49162.1 | |||
| BC057458.1 | AAH57458.1 | |||
| BC057923.1 | AAH57923.1 |
More Information
Set Membership: |
|

Genomic Map