Assay Details
Target Gene Details
Entrez Gene ID: | 11877 |
Gene Name: | armadillo repeat gene deleted in velo-cardio-facial syndrome |
Gene Aliases: |
- |
Location: |
Chr.16:18348182-18407076 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arvcf | NM_001272028.1 | 14 | 2794 | NP_001258957.1 |
| NM_001272029.1 | 14 | 2927 | NP_001258958.1 | |
| NM_001272030.1 | 13 | 2776 | NP_001258959.1 | |
| NM_001272031.1 | 13 | 2537 | NP_001258960.1 | |
| NM_001272032.1 | 12 | 2519 | NP_001258961.1 | |
| NM_033474.3 | 15 | 2945 | NP_258435.2 | |
| AF286212.1 | 14 | 2794 | AAK64214.1 | |
| AF286213.1 | 13 | 2776 | AAK64215.1 | |
| AF286214.1 | 13 | 2446 | AAK64216.1 | |
| AF286215.1 | 12 | 2428 | AAK64217.1 | |
| AJ243418.1 | 13 | 2561 | ||
| AK031855.1 | 8 | 902 | ||
| AK044982.1 | 12 | 2467 | BAC32169.1 | |
| AK051627.1 | 13 | 2493 | BAC34696.1 | |
| AK084886.1 | 14 | 2915 | BAC39302.1 | |
| BC051655.1 | 9 | 942 | AAH51655.1 | |
| BC056980.1 | 15 | 2853 | AAH56980.1 |
More Information
Set Membership: |
|

Genomic Map