Assay Details
Target Gene Details
Entrez Gene ID: | 11877 |
Gene Name: | armadillo repeat gene deleted in velo-cardio-facial syndrome |
Gene Aliases: |
- |
Location: |
Chr.16:18348182-18407076 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arvcf | NM_001272029.1 | NP_001258958.1 | ||
| NM_033474.3 | NP_258435.2 | |||
| AK084886.1 | BAC39302.1 | |||
| BC056980.1 | AAH56980.1 | |||
| BY273838.1 |
More Information
Set Membership: |
|

Genomic Map