Assay Details
Target Gene Details
Entrez Gene ID: | 11877 |
Gene Name: | armadillo repeat gene deleted in velo-cardio-facial syndrome |
Gene Aliases: |
- |
Location: |
Chr.16:18348182-18407076 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arvcf | NM_001272028.1 | 3 | 483 | NP_001258957.1 |
| NM_001272029.1 | 4 | 634 | NP_001258958.1 | |
| NM_001272030.1 | 3 | 483 | NP_001258959.1 | |
| NM_001272031.1 | 2 | 226 | NP_001258960.1 | |
| NM_001272032.1 | 2 | 226 | NP_001258961.1 | |
| NM_033474.3 | 4 | 634 | NP_258435.2 | |
| AF286212.1 | 3 | 483 | AAK64214.1 | |
| AF286213.1 | 3 | 483 | AAK64215.1 | |
| AF286214.1 | 2 | 135 | AAK64216.1 | |
| AF286215.1 | 2 | 135 | AAK64217.1 | |
| AJ243418.1 | 3 | 268 | ||
| AK044982.1 | 2 | 174 | BAC32169.1 | |
| AK051627.1 | 2 | 182 | BAC34696.1 | |
| AK053865.1 | 2 | 207 | ||
| AK084886.1 | 4 | 622 | BAC39302.1 | |
| BC051655.1 | AAH51655.1 | |||
| BC056980.1 | 4 | 542 | AAH56980.1 | |
| BY022048.1 | 2 | 226 |
More Information
Set Membership: |
|

Genomic Map