Assay Details
Target Gene Details
Entrez Gene ID: | 67367 |
Gene Name: | PAX3 and PAX7 binding protein 1 |
Gene Aliases: |
1810007M14Rik, C21orf66, Gcfc, Gcfc1, Pax3/7bp |
Location: |
Chr.16:91014037-91044734 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paxbp1 | NM_026110.2 | NP_080386.3 | ||
| AK007365.1 | BAB24988.2 | |||
| AK079394.1 | ||||
| AY033907.1 | AAK68725.1 | |||
| AY033908.1 | AAK68726.1 | |||
| BC014838.1 | ||||
| BC027145.1 | AAH27145.1 |
More Information
Set Membership: |
|

Genomic Map