Assay Details
Target Gene Details
Entrez Gene ID: | 69457 |
Gene Name: | transmembrane protein 45A2 |
Gene Aliases: |
2310005G13Rik |
Location: |
Chr.16:57036967-57072092 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem45a2 | NM_183281.2 | NP_899104.2 | ||
| AK009173.1 | BAC25243.1 | |||
| AK036849.1 | ||||
| BC150914.1 | ||||
| BC150924.1 | ||||
| BC172084.1 |

Genomic Map