Assay Details
Target Gene Details
Entrez Gene ID: | 20491 |
Gene Name: | src-like adaptor |
Gene Aliases: |
Slap, Slap-1 |
Location: |
Chr.15:66780819-66831829 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sla | NM_001029841.4 | NP_001025012.1 | ||
| NM_009192.3 | NP_033218.1 | |||
| AK036167.1 | BAC29328.1 | |||
| CD578683.1 |
Target Gene Details
Entrez Gene ID: | 21819 |
Gene Name: | thyroglobulin |
Gene Aliases: |
Tgn, cog |
Location: |
Chr.15:66670756-66850721 on Build GRCm38 |
Assay Gene Location: | Within Exon 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tg | NM_009375.2 | 42 | 7336 | NP_033401.2 |
| AB332093.1 | 2 | 153 | ||
| AF076186.1 | 43 | 7314 | AAC32268.1 | |
| AF076187.1 | 43 | 7314 | AAC32269.1 | |
| BC111467.1 | 42 | 7329 | AAI11468.1 | |
| U76389.1 | 42 | 7356 | AAB53204.1 |
More Information
Set Membership: |
|

Genomic Map