Assay Details
Target Gene Details
Entrez Gene ID: | 70508 |
Gene Name: | bobby sox homolog (Drosophila) |
Gene Aliases: |
5530401J07Rik, 5730403O13Rik |
Location: |
Chr.16:50191844-50432461 on Build GRCm38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bbx | NM_027444.3 | NP_081720.2 | ||
| AF454943.1 | AAL58872.1 | |||
| AF454944.1 | AAL58873.1 | |||
| AK029747.1 | BAC26596.1 | |||
| AK041632.1 | BAC31013.1 | |||
| AK211657.1 | ||||
| BC066821.1 | AAH66821.1 |
More Information
Set Membership: |
|

Genomic Map