Assay Details
Target Gene Details
Entrez Gene ID: | 70508 |
Gene Name: | bobby sox homolog (Drosophila) |
Gene Aliases: |
5530401J07Rik, 5730403O13Rik |
Location: |
Chr.16:50191844-50432461 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bbx | NM_027444.3 | 5 | 630 | NP_081720.2 |
| AF276950.1 | 5 | 515 | AAL68986.1 | |
| AF276951.1 | 5 | 602 | AAL68987.1 | |
| AF276952.1 | 2 | 358 | AAL68988.1 | |
| AF454943.1 | 5 | 623 | AAL58872.1 | |
| AF454944.1 | 3 | 495 | AAL58873.1 | |
| AK017487.1 | 5 | 623 | BAB30768.1 | |
| AK029532.1 | 3 | 497 | BAC26500.2 | |
| AK029747.1 | 6 | 744 | BAC26596.1 | |
| AK049516.1 | 5 | 616 | BAC33788.1 | |
| AK050488.1 | 5 | 569 | BAC34285.1 | |
| AK157813.1 | 5 | 609 | BAE34207.1 | |
| AK167724.1 | 5 | 761 | ||
| BC019456.1 | 5 | 705 | ||
| BC024449.1 | 3 | 478 | ||
| BC057869.1 | 5 | 598 | AAH57869.1 | |
| BC066821.1 | AAH66821.1 |
More Information
Set Membership: |
|

Genomic Map