Assay Details
Target Gene Details
Entrez Gene ID: | 224008 |
Gene Name: | scaffolding protein involved in DNA repair |
Gene Aliases: |
2310008H04Rik, Kiaa0146 |
Location: |
Chr.16:15889224-16146851 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spidr | NM_146068.4 | 10 | 1560 | NP_666180.2 |
| AK044836.1 | 10 | 1543 | BAC32113.1 | |
| AK052832.1 | 10 | 1523 | BAC35167.1 | |
| AK075871.1 | 10 | 1525 | BAC36018.1 | |
| AK146053.1 | 10 | 1560 | ||
| AK166448.1 | 5 | 855 | BAE38782.1 | |
| AK170784.1 | 6 | 1192 | BAE42027.1 | |
| AK172902.1 | 10 | 1513 | BAD32180.1 | |
| BC046558.1 | 4 | 564 | AAH46558.1 | |
| BC069859.1 | 1 | 113 | AAH69859.1 | |
| BC089026.1 | 10 | 1497 | AAH89026.1 |
More Information
Set Membership: |
|

Genomic Map