Assay Details
Target Gene Details
Entrez Gene ID: | 67515 |
Gene Name: | tetratricopeptide repeat domain 33 |
Gene Aliases: |
2410099M07Rik, 2900001O04Rik, AI507072, AW538342, Osrf |
Location: |
Chr.15:5185485-5218336 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc33 | NM_026213.3 | NP_080489.1 | ||
| AK010758.1 | ||||
| AK013464.1 | BAB28868.1 | |||
| AK050426.1 | BAC34250.1 | |||
| AK075643.1 | BAC35877.1 | |||
| BC017533.1 | AAH17533.1 | |||
| BC061018.1 | AAH61018.1 |

Genomic Map