Assay Details
Target Gene Details
Entrez Gene ID: | 66736 |
Gene Name: | ER membrane protein complex subunit 2 |
Gene Aliases: |
4921531G14Rik, AV060620, AW209495, Ttc35 |
Location: |
Chr.15:43477214-43527777 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Emc2 | NM_025736.2 | NP_080012.1 | ||
| AF332065.1 | AAK56094.1 | |||
| AF332066.1 | ||||
| AK014989.1 | BAB29657.1 | |||
| AK015968.1 | BAB30057.1 | |||
| AK171442.1 | 2 | 183 | ||
| AK172887.1 | BAD32165.1 | |||
| BC004716.1 | AAH04716.1 |
More Information
Set Membership: |
|

Genomic Map