Assay Details
Target Gene Details
Entrez Gene ID: | 223693 |
Gene Name: | transmembrane protein 184b |
Gene Aliases: |
2610507A11, 4732495E13Rik |
Location: |
Chr.15:79360684-79403303 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem184b | NM_172608.1 | NP_766196.1 | ||
| AK158789.1 | BAE34666.1 | |||
| BY275319.1 |
More Information
Set Membership: |
|

Genomic Map