Assay Details
Target Gene Details
Entrez Gene ID: | 239570 |
Gene Name: | tetratricopeptide repeat domain 38 |
Gene Aliases: |
9030019H09 |
Location: |
Chr.15:85832304-85858822 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc38 | NM_001033337.4 | 4 | 364 | NP_001028509.2 |
| AK078848.1 | 4 | 325 | ||
| AK133636.1 | BAE21760.1 | |||
| BC024550.1 | 4 | 314 | ||
| BC030849.1 | 4 | 327 | ||
| BC132621.1 | 4 | 349 | ||
| BC132623.1 | 4 | 349 | ||
| BY776428.1 |

Genomic Map