Assay Details
Target Gene Details
Entrez Gene ID: | 223697 |
Gene Name: | Sad1 and UNC84 domain containing 2 |
Gene Aliases: |
B230369L08Rik, C030011B15, Unc84b |
Location: |
Chr.15:79724068-79742536 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sun2 | NM_001205345.1 | 3 | 460 | NP_001192274.1 |
| NM_001205346.1 | 3 | 460 | NP_001192275.1 | |
| NM_194342.3 | 3 | 460 | NP_919323.2 | |
| AK046318.1 | 3 | 429 | ||
| AK085196.1 | 3 | 429 | ||
| AK128958.1 | 3 | 454 | BAC87662.1 | |
| AK156246.1 | 3 | 461 | BAE33640.1 | |
| AK171058.1 | 3 | 429 | BAE42217.1 | |
| AK172398.1 | 3 | 461 | ||
| AK172996.1 | 3 | 285 | BAD32274.1 | |
| AY682987.1 | 2 | 226 | AAT90499.1 | |
| BC098208.1 | 3 | 380 | AAH98208.1 |
More Information
Set Membership: |
|

Genomic Map