Assay Details
Target Gene Details
Entrez Gene ID: | 94223 |
Gene Name: | DiGeorge syndrome critical region gene 8 |
Gene Aliases: |
D16H22S1742E, D16H22S788E, D16Wis2, Gy1, N41, Vo59c07, mir-1306 |
Location: |
Chr.16:18253948-18289252 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgcr8 | NM_033324.2 | NP_201581.2 | ||
| AB086857.1 | BAD06468.1 | |||
| AK086233.1 | ||||
| AK161410.1 | ||||
| AK180426.1 | ||||
| BC062919.1 | AAH62919.1 | |||
| BC066118.1 | AAH66118.1 |
More Information
Set Membership: |
|

Genomic Map