Assay Details
Target Gene Details
Entrez Gene ID: | 14167 |
Gene Name: | fibroblast growth factor 12 |
Gene Aliases: |
AV114868, B230343J05Rik, FGF-12, FHF-1, Fhf1 |
Location: |
Chr.16:28157783-28753243 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgf12 | NM_001276420.1 | 1 | 924 | NP_001263349.1 |
| AF020738.1 | 1 | 120 | AAB71607.1 | |
| AF020739.1 | AAB71608.1 | |||
| AK011712.2 | ||||
| AK018774.1 | ||||
| AK034335.1 | 1 | 77 | BAC28679.1 | |
| AK148215.1 | 1 | 924 | BAE28417.1 | |
| BB611389.1 | ||||
| BC030485.1 | AAH30485.1 | |||
| CX239635.1 | ||||
| U66201.1 | AAB18917.1 |
More Information
Set Membership: |
|

Genomic Map