Assay Details
Target Gene Details
Entrez Gene ID: | 666173 |
Gene Name: | vacuolar protein sorting 13B (yeast) |
Gene Aliases: |
1810042B05Rik, 2310042E16Rik, 4732488H20, C330002D13Rik, C87206, Coh1, D230005K13, mKIAA0532, mKIAA5032 |
Location: |
Chr.15:35369882-35931229 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps13b | NM_177151.3 | 16 | 2528 | NP_796125.2 |
More Information
Set Membership: |
|

Genomic Map