Assay Details
Target Gene Details
Entrez Gene ID: | 11504 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 1 |
Gene Aliases: |
ADAM-TS1, ADAMTS, ADAMTS-1, C3-C5, METH-1, METH1 |
Location: |
Chr.16:85793827-85803115 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamts1 | NM_009621.4 | NP_033751.3 | ||
| AK156830.1 | BAE33869.1 | |||
| AK161409.1 | BAE36379.1 | |||
| AK163622.1 | BAE37426.1 | |||
| BC003269.1 | ||||
| BC040382.1 | AAH40382.1 | |||
| BC050834.1 | AAH50834.1 | |||
| D67076.1 | BAA11088.1 |
More Information
Set Membership: |
|

Genomic Map