Assay Details
Target Gene Details
Entrez Gene ID: | 22762 |
Gene Name: | zinc finger protein, multitype 2 |
Gene Aliases: |
B330005D23Rik, FOG-2, FOG2 |
Location: |
Chr.15:40655042-41104592 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfpm2 | NM_011766.5 | 7 | 1391 | NP_035896.1 |
| AF107306.1 | 7 | 912 | AAD12182.1 | |
| AF118845.1 | 7 | 1161 | AAD17251.1 | |
| AF125166.1 | 7 | 1392 | AAD33250.1 | |
| BC059241.1 | 7 | 1291 | AAH59241.1 | |
| EF514219.1 | 7 | 2214 |
More Information
Set Membership: |
|

Genomic Map