Assay Details
Target Gene Details
Entrez Gene ID: | 13383 |
Gene Name: | discs, large homolog 1 (Drosophila) |
Gene Aliases: |
B130052P05Rik, Dlgh1, E-dlg/SAP97, SAP-97, SAP97, mKIAA4187 |
Location: |
Chr.16:31663443-31873356 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlg1 | NM_001252433.1 | NP_001239362.1 | ||
| NM_001252434.1 | NP_001239363.1 | |||
| NM_001252435.1 | NP_001239364.1 | |||
| NM_001252436.1 | NP_001239365.1 | |||
| NM_007862.3 | NP_031888.2 | |||
| NR_045516.1 | ||||
| AK048240.1 | 1 | 1047 | ||
| AK142893.1 | ||||
| AK143780.1 | BAE25536.1 | |||
| AK160918.1 | ||||
| AK220189.1 | BAD90374.1 | |||
| AY159380.1 | AAN87264.1 | |||
| BC047142.1 | AAH47142.1 | |||
| BC057118.1 | AAH57118.1 | |||
| U93309.1 | AAC31653.1 |
More Information
Set Membership: |
|

Genomic Map