Assay Details
Target Gene Details
Entrez Gene ID: | 13837 |
Gene Name: | Eph receptor A3 |
Gene Aliases: |
AW492086, Cek4, ETK1, End3, Hek, Hek4, Mek4, Tyro4 |
Location: |
Chr.16:63545218-63864157 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epha3 | NM_010140.3 | 16 | 2922 | NP_034270.1 |
| AK045219.1 | 16 | 2923 | BAC32266.1 | |
| AK084921.1 | 16 | 2902 | BAC39310.1 | |
| AK132335.1 | 16 | 2871 | BAE21112.1 | |
| BC138641.1 | 16 | 2845 | ||
| BC138643.1 | 16 | 2845 | ||
| BC145349.1 | 16 | 2842 | ||
| M68513.1 | 16 | 2897 | AAA39521.1 |
More Information
Set Membership: |
|

Genomic Map