Assay Details
Target Gene Details
Entrez Gene ID: | 13837 |
Gene Name: | Eph receptor A3 |
Gene Aliases: |
AW492086, Cek4, ETK1, End3, Hek, Hek4, Mek4, Tyro4 |
Location: |
Chr.16:63545218-63864157 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epha3 | NM_010140.3 | 4 | 1040 | NP_034270.1 |
| AK041935.1 | 4 | 1041 | BAC31104.1 | |
| AK045219.1 | 4 | 1041 | BAC32266.1 | |
| AK084921.1 | 4 | 1020 | BAC39310.1 | |
| AK132335.1 | 4 | 989 | BAE21112.1 | |
| BC138641.1 | 4 | 963 | ||
| BC138643.1 | 4 | 963 | ||
| BC145349.1 | 4 | 963 | ||
| M68513.1 | 4 | 1015 | AAA39521.1 | |
| M68515.1 | 4 | 1015 | AAA39522.1 |
More Information
Set Membership: |
|

Genomic Map