Assay Details
Target Gene Details
Entrez Gene ID: | 13840 |
Gene Name: | Eph receptor A6 |
Gene Aliases: |
Ehk2, Hek12, m-ehk2 |
Location: |
Chr.16:59653061-60606484 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epha6 | NM_007938.2 | 11 | 2536 | NP_031964.2 |
| AK032436.1 | 7 | 781 | BAC27868.1 | |
| AK077099.1 | 5 | 546 | BAE43358.1 | |
| BC141090.1 | 11 | 2052 | ||
| U58332.1 | 11 | 2252 | AAB53836.1 |
More Information
Set Membership: |
|

Genomic Map