Assay Details
Target Gene Details
Entrez Gene ID: | 72960 |
Gene Name: | DNA topoisomerase 1, mitochondrial |
Gene Aliases: |
2900052H09Rik |
Location: |
Chr.15:75657029-75678822 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Top1mt | NM_028404.2 | NP_082680.1 | ||
| AF362952.1 | AAM00270.1 | |||
| AK147119.1 | BAE27691.1 | |||
| BC138582.1 | ||||
| BC138583.1 | ||||
| BC145336.1 |
More Information
Set Membership: |
|

Genomic Map